Tetrabutylammonium hydroxide 40% in aqueous solution, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

86880-100ML 86880-2.5L 86880-500ML
SIAL86880-100MLEA 61.2 GBP
SIAL86880-100ML SIAL86880-2.5L SIAL86880-500ML
Tetrabutylammonium hydroxide 40% in aqueous solution, Sigma-Aldrich®
Tetrabutylammonium hydroxide
Formula: C₁₆H₃₇NO
Storage Temperature: Refrigerator
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR