Stearyl stearate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

85775-1G 85775-5G
SIAL85775-1GEA 268 GBP
SIAL85775-1G SIAL85775-5G
Stearyl stearate, Sigma-Aldrich®
Stearyl stearate
Formula: C₃₆H₇₂O₂
Boiling Pt: ∼175 °C (1013 hPa)
Melting Pt: 64 °C
Flash Pt: >160 °C
Storage Temperature: Ambient
MDL Number: MFCD00056225
CAS Number: 2778-96-3
EINECS: 220-476-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR