Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

85590-25G 85590-5G
SIAL85590-25GEA 788 GBP
SIAL85590-25G SIAL85590-5G
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine), Sigma-Aldrich®
Spermine (N,N'-Bis(3-aminopropyl)-1,4-butanediamine)
Formula: C₁₀H₂₆N₄
MDL Number: MFCD00008215
CAS Number: 71-44-3
EINECS: 200-754-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR