D-(+)-Glucurono-6,3-lactone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (R)-2-((2S,3R,4S)-3,4-Dihydroxy-5-oxotetrahydrofuran-2-yl)-2-hydroxyacetaldehyde, D-(+)-Glucuronolactone

M-Clarity™ quality level = MQ200

851450-100G-A 851450-1KG-A
SIAL851450-100G-AEA 80.5 GBP
SIAL851450-100G-A SIAL851450-1KG-A
D-(+)-Glucurono-6,3-lactone, Sigma-Aldrich®
D(+)-Glucurono-6,3-lactone
Formula: C₆H₈O₆
MDL Number: MFCD00135622
CAS Number: 32449-92-6
EINECS: 251-053-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR