2-Pyrrolidone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Pyrrolidin-2-one

M-Clarity™ quality level = MQ200

83300-1L 83300-250ML
SIAL83300-1LEA 121 GBP
SIAL83300-1L SIAL83300-250ML
2-Pyrrolidone, Sigma-Aldrich®
2-Pyrrolidone
Formula: C₄H₇NO
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 24…26 °C
Density: 1.1115 g/cm³ (20 °C)
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD00005270
CAS Number: 616-45-5
EINECS: 210-483-1
Merck Index: 12,08200

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR