Di-n-octyl phthalate 98,0 GC_ASSAY_METHOD, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

80153-100ML
SIAL80153-100MLEA 233 GBP
SIAL80153-100ML
Di-n-octyl phthalate 98,0 GC_ASSAY_METHOD, Sigma-Aldrich®
Di-n-octyl phthalate
Formula: C₂₄H₃₈O₄
MW: 390.56 g/mol
Density: 0.980 g/mL (20 °C) (lit.)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00015292
CAS Number: 117-84-0
EINECS: 204-214-7
UN: 3082
ADR: 9 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR