Phloroglucinol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1,3,5-Trihydroxybenzene

79330-100G 79330-25G
SIAL79330-100GEA 91.3 GBP
SIAL79330-100G SIAL79330-25G
Phloroglucinol, Sigma-Aldrich®
Phloroglucinol
Formula: C₆H₆O₃
Boiling Pt: 400 °C (1013 hPa)
Melting Pt: 217…220 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002286
CAS Number: 108-73-6
EINECS: 203-611-2
Merck Index: 12,07482

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR