4'-Ethoxyacetanilide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Acetphenetidin, Acetophenetidin, Phenacetin, N-(4-Ethoxyphenyl)acetamide, 4-Acetophenetidide

77440-1KG 77440-250G 77440-50G
SIAL77440-1KGEA 150 GBP
SIAL77440-1KG SIAL77440-250G SIAL77440-50G
4'-Ethoxyacetanilide, Sigma-Aldrich®
4'-Ethoxyacetanilide
Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD00009094
CAS Number: 62-44-2
EINECS: 200-533-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR