Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite ≥95%

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Bis(2-cyanoethyl) diisopropylphosphoramidite, Bis(2-cyanoethoxy)(diisopropylamino)phosphine, Bis(2-cyanoethoxy)-N,N-diisopropylaminophosphine

766305-1G 766305-5G
SIAL766305-1GEA 175 GBP
SIAL766305-1G SIAL766305-5G
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite ≥95%
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite

Useful phosphorylating reagent used in oligonucleotide synthesis for adding a terminal phosphate group to the 3′ or 5′ hydroxyl.

Formula: C₁₂H₂₂N₃O₂P
MDL Number: MFCD00797597
CAS Number: 102690-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR