n-Pentadecane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: N-Pentadecane, Pentadecane

76510-100ML 76510-25ML
SIAL76510-100MLEA 148 GBP
SIAL76510-100ML SIAL76510-25ML
n-Pentadecane, Sigma-Aldrich®
n-Pentadecane
Formula: H₃C(CH₂)₁₃CH₃
MW: 212.42 g/mol
Boiling Pt: 271
Melting Pt: 13…15 °C
Density: 0.768 g/cm³ (20 °C)
Flash Pt: 132
Storage Temperature: Ambient
MDL Number: MFCD00008990
CAS Number: 629-62-9
EINECS: 211-098-1
UN: 3295
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR