Guanosine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

M-Clarity™ quality level = MQ200

75903-100G
SIAL75903-100GEA 245 GBP
SIAL75903-100G
Guanosine, Sigma-Aldrich®
Guanosine
Formula: C₁₀H₁₃N₅O₅
Storage Temperature: Ambient
MDL Number: MFCD00010182
CAS Number: 118-00-3
EINECS: 204-227-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR