Sodium dithionite, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Sodium hydrosulphite

71699-1KG 71699-250G 71699-50G
SIAL71699-1KGEA 272 GBP
SIAL71699-1KG SIAL71699-250G SIAL71699-50G
Sodium dithionite, Sigma-Aldrich®
Sodium dithionite
Formula: Na₂S₂O₄
MW: 174.11 g/mol
Melting Pt: 100
Density: 2.19 g/cm³ (20 °C)
Flash Pt: 101
Storage Temperature: Ambient
MDL Number: MFCD00011640
CAS Number: 7775-14-6
EINECS: 231-890-0
UN: 1384
ADR: 4.2 II
Merck Index: 13,08700

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR