Coated slides
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
703176-10PAK
SIAL703176-10PAKEA 95.9 GBP
SIAL703176-10PAK
Coated slides
Slides Adhesive Slides
Indium tin oxide coated glass slides.
  • Good resistivity of 70-100 Ω/sq
  • Unpolished surface with 86 % transmittance
Order Now
SPECIFICATIONS
Environmentally Preferable