(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (R)-(+)-BINAP, (R)-BINAP, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

M-Clarity™ quality level = MQ100

693065-100MG 693065-500MG
SIAL693065-100MGEA 38.5 GBP
SIAL693065-100MG SIAL693065-500MG
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, Sigma-Aldrich®
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formula: C₄₄H₃₂P₂
Melting Pt: 239…242 °C
MDL Number: MFCD00010805
CAS Number: 76189-55-4
Merck Index: 13,01223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR