1-Hexadecanethiol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: n-Hexadecyl mercaptan, 1-Mercaptohexadecane, Hexadecane-1-thiol, n-Hexadecanethiol

M-Clarity™ quality level = MQ100

674516-500MG
SIAL674516-500MGEA 500 GBP
SIAL674516-500MG
1-Hexadecanethiol, Sigma-Aldrich®
1-Hexadecanethiol
Formula: C₁₆H₃₄S
MW: 258.51 g/mol
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 18…20 °C
Density: 0.8556 g/cm³ (20 °C)
Flash Pt: 99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011677
CAS Number: 2917-26-2
EINECS: 220-846-6
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR