Potassium-tert-butoxide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Potassium-2-methylpropan-2-olate, Potassium tert-butylate, Potassium tert-butanolate, Potassium t-butoxide

M-Clarity™ quality level = MQ100

659878-25G 659878-5G
SIAL659878-25GEA 313 GBP
SIAL659878-25G SIAL659878-5G
Potassium-tert-butoxide, Sigma-Aldrich®
Potassium-tert-butoxide
Formula: C₄H₉KO
Melting Pt: 256…258 °C
MDL Number: MFCD00012162
CAS Number: 865-47-4
EINECS: 212-740-3
UN: 3206
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR