Bismuth(III) bromide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ100

654981-1G 654981-5G
SIAL654981-1GEA 93.6 GBP
SIAL654981-1G SIAL654981-5G
Bismuth(III) bromide, Sigma-Aldrich®
Bismuth (III) bromide
Formula: BiBr₃
Boiling Pt: 278 °C (11 mmHg)
Melting Pt: 218 °C
Density: 5.72 g/cm³ (20 °C)
MDL Number: MFCD00014195
CAS Number: 7787-58-8
EINECS: 232-121-1
UN: 3260
ADR: 8,III
Merck Index: 12,01299

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR