2,4-Dichloro-5-fluoropyrimidine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 5-Fluoro-2,4-dichloropyrimidine

M-Clarity™ quality level = MQ100

653233-1G
SIAL653233-1GEA 38.8 GBP
SIAL653233-1G
2,4-Dichloro-5-fluoropyrimidine, Sigma-Aldrich®
2,4-Dichloro-5-fluoropyrimidine
Formula: C₄HCl₂FN₂
MW: 166.97 g/mol
MDL Number: MFCD00233551
CAS Number: 2927-71-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR