2-(Methylthio)ethylamine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-(Methylthio)ethanamine

M-Clarity™ quality level = MQ100

632929-1G 632929-5G
SIAL632929-1GEA 124 GBP
SIAL632929-1G SIAL632929-5G
2-(Methylthio)ethylamine, Sigma-Aldrich®
2-(Methylthio)ethylamine
Formula: C₃H₉NS
MDL Number: MFCD00014825
CAS Number: 18542-42-2
EINECS: 242-412-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR