Ethyl laurate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

61630-100ML
SIAL61630-100MLEA 74.3 GBP
SIAL61630-100ML
Ethyl laurate, Sigma-Aldrich®
Ethyl laurate
Formula: C₁₄H₂₈O₂
Boiling Pt: 269 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00015065
CAS Number: 106-33-2
EINECS: 203-386-0
Merck Index: 13,03851

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR