Silicon carbide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ100

594911-100G 594911-250G
SIAL594911-100GEA 1130 GBP
SIAL594911-100G SIAL594911-250G
Silicon carbide, Sigma-Aldrich®
Silicon carbide
Formula: SiC
Melting Pt: 2700 °C
Density: 3.22 g/cm³ (25 °C)
MDL Number: MFCD00049531
CAS Number: 409-21-2
EINECS: 206-991-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd532b391b9c28edcb8adac5fe5ed958ccaa487&vid=d39ce1cfd6c1a5ca44c7d11b656758cbfaed190d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR