Hafnium carbide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

594636-25G
SIAL594636-25GEA 419 GBP
SIAL594636-25G
Hafnium carbide, Sigma-Aldrich®
Hafnium carbide
Formula: CHf
MDL Number: MFCD00016126
CAS Number: 12069-85-1
EINECS: 235-114-1
UN: 3178
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR