Chloroacetamidine hydrochloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-Chloroacetimidamide hydrochloride, 2-Chloroacetamidine hydrochloride

M-Clarity™ quality level = MQ100

591475-5G
SIAL591475-5GEA 73 GBP
SIAL591475-5G
Chloroacetamidine hydrochloride, Sigma-Aldrich®
Chloroacetamidine hydrochloride
Formula: C₂H₆Cl₂N₂
MDL Number: MFCD00053013
CAS Number: 10300-69-3
EINECS: 233-675-7
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR