3-(Boc-amino)phenylboronic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3-Aminobenzeneboronic acid, N-BOC protected, 3-(N-Boc-amino)phenylboronic acid, (3-((tert-Butoxycarbonyl)amino)phenyl)boronic acid, 3-Boronoaniline, N-BOC protected, 3-[(tert-Butoxycarbonyl)amino]benzeneboronic acid, 3-(Boc-amino)benzeneboronic acid

M-Clarity™ quality level = MQ100

578851-5G
SIAL578851-5GEA 163 GBP
SIAL578851-5G
3-(Boc-amino)phenylboronic acid, Sigma-Aldrich®
3-(Boc-amino)phenylboronic acid
Formula: C₁₁H₁₆BNO₄
MW: 237.06 g/mol
MDL Number: MFCD03411945
CAS Number: 380430-68-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR