Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

56485-250MG 56485-1G 56485-5G
SIAL56485-250MGEA 155 GBP
SIAL56485-250MG SIAL56485-1G SIAL56485-5G
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt), Sigma-Aldrich®
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt)

N-Hydroxysulfosuccinimide sodium salt is a water-soluble sulfonated analog of N-hydroxysuccinimide. It can undergo a coupling reaction with carboxylic acids in the presence of carbodiimides such as 1-ethyl-3-(3-dimethylaminopropyl)carbodiimide(EDC) to form hydrophilic N-hydroxysulfosuccinimide active esters. These esters are useful in protein cross-linking experiments.

Formula: C₄H₄NNaO₆S
Storage Temperature: Freezer
MDL Number: MFCD00043100
CAS Number: 106627-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR