1-Iodo-2-(trifluoromethoxy)benzene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

554391-5G
SIAL554391-5GEA 97.2 GBP
SIAL554391-5G
1-Iodo-2-(trifluoromethoxy)benzene, Sigma-Aldrich®
1-Iodo-2-(trifluoromethoxy)benzene
Formula: C₇H₄F₃IO
MDL Number: MFCD00042410
CAS Number: 175278-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR