2-Chlorobenzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-Chlorobenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ100

546925-25G 546925-5G
SIAL546925-25GEA 179 GBP
SIAL546925-25G SIAL546925-5G
2-Chlorobenzenesulphonyl chloride, Sigma-Aldrich®
2-Chlorobenzenesulphonyl chloride
Formula: C₆H₄Cl₂O₂S
Boiling Pt: 88…92 °C (0.1 mmHg)
Melting Pt: 28 °C
Density: 1.548 g/cm³ (20 °C)
MDL Number: MFCD00051520
CAS Number: 2905-23-9
EINECS: 220-807-3
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR