Ytterbium triacetate hydrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Ytterbium(III) acetate hydrate

M-Clarity™ quality level = MQ100

544973-25G 544973-5G
SIAL544973-25GEA 366 GBP
SIAL544973-25G SIAL544973-5G
Ytterbium triacetate hydrate, Sigma-Aldrich®
Ytterbium triacetate hydrate
Formula: C₆H₉O₆Yb·H₂O
MDL Number: MFCD00150127
CAS Number: 15280-58-7
EINECS: 244-137-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR