Zinc oxide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ200

544906-10G 544906-50G
SIAL544906-10GEA 40.6 GBP
SIAL544906-10G SIAL544906-50G
Zinc oxide, Sigma-Aldrich®
Zinc oxide
Formula: ZnO
Melting Pt: 1975 °C
Density: 5.61 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR