1-Dodecene-[1,2-¹³C₂] (99% ¹³C ), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

491209-100MG
SIAL491209-100MGEA 2420 GBP
SIAL491209-100MG
1-Dodecene-[1,2-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
1-Dodecene-[1,2-13C2]
Formula: C₁₂H₂₄
Storage Temperature: Ambient
CAS Number: 198332-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR