Ammonium nitrate-[¹⁵N₂] (5% ¹⁵N), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Ammonium nitrate -15N2, Ammonium-15N nitrate-15N, Norway saltpeter-15N2

M-Clarity™ quality level = MQ200

488070-100G
SIAL488070-100GEA 585 GBP
SIAL488070-100G
Ammonium nitrate-[¹⁵N₂] (5% ¹⁵N), Sigma-Aldrich®
Ammonium nitrate-[15N2]
Formula: H₄N₂O₃
MDL Number: MFCD00082562
CAS Number: 43086-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR