Silver trifluoromethanesulphonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Silver trifluoromethanesulphonate, Silver triflate, Trifluoromethanesulphonic acid silver salt

M-Clarity™ quality level = MQ100

483346-1G 483346-5G
SIAL483346-1GEA 62.9 GBP
SIAL483346-1G SIAL483346-5G
Silver trifluoromethanesulphonate, Sigma-Aldrich®
Silver trifluoromethanesulphonate
Formula: CAgF₃O₃S
Melting Pt: 356 °C
MDL Number: MFCD00013226
CAS Number: 2923-28-6
EINECS: 220-882-2
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR