4-Phenoxyphenylboronic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (4-Phenoxyphenyl)boronic acid, 4-Phenoxybenzeneboronic acid

M-Clarity™ quality level = MQ100

480142-5G
SIAL480142-5GEA 75 GBP
SIAL480142-5G
4-Phenoxyphenylboronic acid, Sigma-Aldrich®
4-Phenoxyphenylboronic acid
Formula: C₁₂H₁₁BO₃
MW: 214.03 g/mol
MDL Number: MFCD00093312
CAS Number: 51067-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR