4-Bromobenzylamine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ100

479152-1G
SIAL479152-1GEA 148 GBP
SIAL479152-1G
4-Bromobenzylamine, Sigma-Aldrich®
4-Bromobenzylamine
Formula: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD00047931
CAS Number: 3959-07-7
EINECS: 223-559-4
UN: 3259
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR