1-Iodo-3-methylbutane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3-Methylbutyliodide

MQ100

476447-10G
SIAL476447-10GEA 62.3 GBP
SIAL476447-10G
1-Iodo-3-methylbutane, Sigma-Aldrich®
1-Iodo-3-methylbutane
Formula: C₅H₁₁I
MW: 198.05 g/mol
Boiling Pt: 146…148 °C (1013 hPa)
Density: 1.515 g/cm³ (20 °C)
MDL Number: MFCD00039414
CAS Number: 541-28-6
EINECS: 208-773-8
UN: 1993
ADR: 3 III
Merck Index: 13,05136

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR