Potassium tetrahydroborate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

455571-100G 455571-500G
SIAL455571-100GEA 127 GBP
SIAL455571-100G SIAL455571-500G
Potassium tetrahydroborate, Sigma-Aldrich®
Potassium tetrahydroborate
Formula: H₄BK
Melting Pt: 500 °C
Density: 1.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011396
CAS Number: 13762-51-1
EINECS: 237-360-5
UN: 1870
ADR: 4.3,I
Merck Index: 12,07777

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR