Ethanol-[D₁], Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Hydroxy ethane-D1, Ethyl alcohol-D1, Ethanol-[²H₁], Methyl carbinol-D1

M-Clarity™ quality level = MQ100

452556-100ML 452556-25ML
SIAL452556-100MLEA 198 GBP
SIAL452556-100ML SIAL452556-25ML
Ethanol-[D₁], Sigma-Aldrich®
Ethanol-[D1]
Formula: C₂H₅DO
Boiling Pt: 78 °C (1013 hPa)
Melting Pt: –114.5 °C
Density: 0.826 g/cm³ (20 °C)
CAS Number: 925-93-9
EINECS: 213-128-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR