Yttrium(III) chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ100

450103-25G 450103-5G
SIAL450103-25GEA 378 GBP
SIAL450103-25G SIAL450103-5G
Yttrium(III) chloride, Sigma-Aldrich®
Yttrium (III) chloride
Formula: Cl₃Y
Boiling Pt: 1488 °C (1013 hPa)
Melting Pt: 703 °C
Density: 2.52 g/cm³ (700 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011470
CAS Number: 10361-92-9
EINECS: 233-801-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR