3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3-(1,1,1,5,5,5-Hexamethyl-3-((trimethylsilyl)oxy)trisiloxan-3-yl)propyl methacrylate, 3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane

M-Clarity™ quality level = MQ100

446130-25ML
SIAL446130-25MLEA 142 GBP
SIAL446130-25ML
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate, Sigma-Aldrich®
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate
Formula: C₁₆H₃₈O₅Si₄
MDL Number: MFCD00053871
CAS Number: 17096-07-0
EINECS: 241-165-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR