HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: HATU, 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxide, hexafluorophosphate

M-Clarity™ quality level = MQ200

445460-1G 445460-25G 445460-5G
SIAL445460-1GEA 74.5 GBP
SIAL445460-1G SIAL445460-25G SIAL445460-5G
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate), Sigma-Aldrich®
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
Formula: C₁₀H₁₅F₆N₆OP
MDL Number: MFCD00274639
CAS Number: 148893-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR