4-Fluoro-2-(trifluoromethyl)benzaldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


MQ200

436097-1G 436097-5G
SIAL436097-1GEA 54.3 GBP
SIAL436097-1G SIAL436097-5G
4-Fluoro-2-(trifluoromethyl)benzaldehyde, Sigma-Aldrich®
4-Fluoro-2-(trifluoromethyl)benzaldehyde
Formula: C₈H₄F₄O
MW: 192.11 g/mol
Boiling Pt: 114
Density: 1.4 g/cm³ (20 °C)
MDL Number: MFCD00061313
CAS Number: 90176-80-0
UN: 1989
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR