Acetone-[D₆], Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Propanone-D6, DMK-D6, Hexadeuteroacetone, Dimethyl ketone-D6, Acetone-[²H₆], Acetone-d6

MQ100

434531-25G 434531-50G
SIAL434531-25GEA 185 GBP
SIAL434531-25G SIAL434531-50G
Acetone-[D₆], Sigma-Aldrich®
Acetone-[D6]
Formula: D₃CC(O)CD₃
MW: 64.03 g/mol
Boiling Pt: 55.5
Melting Pt: –95.4 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: –19 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00044635
CAS Number: 666-52-4
EINECS: 211-563-9
UN: 1090
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR