Toluene-[D₈], Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Phenylmethane-D8, Methylbenzene-D8, Toluene-[²H₈]

M-Clarity™ quality level = MQ100

434388-10G 434388-25G 434388-5G
SIAL434388-10GEA 156 GBP
SIAL434388-10G SIAL434388-25G SIAL434388-5G
Toluene-[D₈], Sigma-Aldrich®
Toluene-[D8]
Formula: C₆D₅CD₃
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –85 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 4 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00044638
CAS Number: 2037-26-5
EINECS: 218-009-5
UN: 1294
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR