Potassium iodide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Dipotassium diiodide, Kali iodide

MQ100

429422-25G 429422-5G
SIAL429422-25GEA 123 GBP
SIAL429422-25G SIAL429422-5G
Potassium iodide, Sigma-Aldrich®
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686
Density: 3.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR