10-Methylphenothiazine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

425346-5G
SIAL425346-5GEA 109 GBP
SIAL425346-5G
10-Methylphenothiazine, Sigma-Aldrich®
10-Methylphenothiazine
Formula: C₁₃H₁₁NS
MW: 213.3 g/mol
Boiling Pt: 342 °C (1013 hPa)
Melting Pt: 99…100 °C
MDL Number: MFCD00041836
CAS Number: 1207-72-3
EINECS: 214-896-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR