1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 1H,1H,10H,10H-Perfluoro-1,10-decandiol, 1H,1h,10h,10h-perfluoro-1,10-decanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1,10-decandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1,10-decanediol, 1H,1H,10H,10H-Perfluoro-1,10-decanediol, 1H,1H,10H,10H-Perfluorodecane-1,10-diol

M-Clarity™ quality level = MQ100

406600-5G
SIAL406600-5GEA 66.5 GBP
SIAL406600-5G
1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol, Sigma-Aldrich®
1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol
Formula: C₁₀H₆F₁₆O₂
MW: 462.13 g/mol
MDL Number: MFCD00192200
CAS Number: 754-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR