Tetrafluoroboric acid diethyl ether complex (51 - 57% HBF₄) (w/w), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

400068-100ML 400068-25ML 400068-400ML
SIAL400068-100MLEA 140 GBP
SIAL400068-100ML SIAL400068-25ML SIAL400068-400ML
Tetrafluoroboric acid diethyl ether complex (51 - 57% HBF₄) (w/w), Sigma-Aldrich®
Tetrafluoroboric acid-diethyl ether complex (50-55% tetrafluoroboric acid in diethyl ether)
Formula: C₄H₁₁BF₄O
MDL Number: MFCD00085352
CAS Number: 67969-82-8
EINECS: 267-983-8
UN: 3129
ADR: 4.3 (8),II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR