Cyclohexanone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

398241-25ML 398241-2L 398241-500ML
SIAL398241-25MLEA 61.9 GBP
SIAL398241-25ML SIAL398241-2L SIAL398241-500ML
Cyclohexanone, Sigma-Aldrich®
Cyclohexanone
Formula: C₆H₁₀(=O)
Boiling Pt: 155.6 °C (1013 hPa)
Melting Pt: –31 °C
Density: 0.9478 g/cm³ (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00001625
CAS Number: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3,III
REACH: 01-2119453616-35
Merck Index: 13,02755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8701dac7edb70a452483cd4f1bd716ca29ba9aa&vid=0360daffe7606174162954ba58a6ba2b78e28dfb&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR