Potassium hexachloroplatinate(IV), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Dipotassium hexachloroplatinate

M-Clarity™ quality level = MQ100

379859-1G 379859-5G
SIAL379859-1GEA 185 GBP
SIAL379859-1G SIAL379859-5G
Potassium hexachloroplatinate(IV), Sigma-Aldrich®
Potassium hexachloroplatinate(IV)
Formula: K₂PtCl₆
Melting Pt: 250 °C
Density: 3.344 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011389
CAS Number: 16921-30-5
EINECS: 240-979-3
UN: 3290
ADR: 6.1,II
Merck Index: 13,07720

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR