[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

379670-1G 379670-25G 379670-5G
SIAL379670-1GEA 92.8 GBP
SIAL379670-1G SIAL379670-25G SIAL379670-5G
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex, Sigma-Aldrich®
[1,1'-Bis(diphenylphosphino)ferrocene]palladium (II) chloride dichloromethane complex
Formula: C₃₅H₃₀Cl₄FeP₂Pd
MDL Number: MFCD00792899
CAS Number: 95464-05-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR